Tacrolimus ≥98%, polvo cristalino

Proveedor: ENZO LIFE SCIENCES
Peligro

ALX380008M001 ALX380008M005 ALX380008M025
ENZOALX380008M001EA 55 EUR
ENZOALX380008M001 ENZOALX380008M005 ENZOALX380008M025
Tacrolimus ≥98%, polvo cristalino
Tacrolimus

Immunosuppressant.


Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2. Shown to inhibit the activity of FK506 binding protein and to inhibit the Ca²⁺-dependent phosphatase and protein phosphatase 2B (PP2B; calcineurin). Activates NF-κB through phosphorylation and degradation of IκBα.


Soluble in DMSO, 100% ethanol, methanol, ethyl acetate or acetone; insoluble in water.

Formula: C₄₄H₆₉NO₁₂
Temperatura de almacenaje: Congelador
Número MDL: MFCD00869853
Núm. CAS: 104987-11-3
UN: 2811
ADR: 6.1,III
Índice Merck: 14,09025

Order Now

Resultados test especificaciones

Purity ≥98% (Assay)
Source Isolated from Streptomyces tsukubaensi

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR