Bopindolol malonate ≥97% (by HPLC)

Proveedor: Enzo Life Sciences

BMLAR1000100 BMLAR1000500
ENZOBMLAR1000100EA 84.7 EUR
ENZOBMLAR1000100 ENZOBMLAR1000500
Bopindolol malonate ≥97% (by HPLC)
Bopindolol malonate

Adrenergic receptor ligand.


A slowly dissociating β1-adrenergic receptor antagonist. Antihypertensive. Antioxidant (IC₅₀ = 1.8 µM) and free radical scavenging activity.


Soluble in water at 3 mg/ml.

Formula: C₂₃H₂₈N₂O₃·C₃H₄O₄
Número MDL: MFCD00798228
Núm. CAS: 62658-64-4

Order Now

Resultados test especificaciones

Purity  ≥97% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR