Anti-Llama IgG Goat Ployclonal Antibody (CF®583R)

Proveedor: Biotium
20902-500UL 20902-50UL
20902-500ULEA 201 EUR
20902-500UL 20902-50UL
Anti-Llama IgG Goat Ployclonal Antibody (CF®583R)
Anticuerpos
This is a goat anti-llama IgG (H+L) secondary antibody labeled with our superior CF® Dyes, HRP, or DNP.

  • Available in 14 bright and photostable CF® Dyes, HRP, or DNP
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Conjugates of blue fluorescent dyes like CF®405S are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colours.

Type: Secondary
Antigen: Llama IgG
Clonality: Ployclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Llama
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR