Anti-IgG Llama Polyclonal Antibody (CF®583R)

Proveedor: Biotium
20908-50UL 20908-500UL
20908-50ULEA 87.4 EUR
20908-50UL 20908-500UL
Anti-IgG Llama Polyclonal Antibody (CF®583R)
Anticuerpos
Llama anti-mouse IgG (H L) secondary antibody labelled with superior CF®583R dye.

  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR