Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 555)
Proveedor: AAT BIOQUEST
10081090 10081091
AATB10081090EA 371 EUR
AATB10081090 AATB10081091
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 555)
Anticuerpos
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Precaución: For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 555
Epitope:
Host: Mouse
Isotype:
Reactivity: Human