Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 560)

Proveedor: AAT Bioquest
101610A0 101610A1
AATB101610A0EA 371 EUR
AATB101610A0 AATB101610A1
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 560)
Anticuerpos
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: iFluor® 560
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR