Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Blue 570)
Proveedor: AAT BIOQUEST
100810T0 100810T1
AATB100810T0EA 371 EUR
AATB100810T0 AATB100810T1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Blue 570)
Anticuerpos
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Precaución: For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human