Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Red 700)
Proveedor: AAT BIOQUEST
100810V0 100810V1
AATB100810V0EA 371 EUR
AATB100810V0 AATB100810V1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Red 700)
Anticuerpos
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Precaución: For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: mFluor™ Red 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human