Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Violet 450)

Proveedor: AAT BIOQUEST
100810Z0 100810Z1
AATB100810Z0EA 371 EUR
AATB100810Z0 AATB100810Z1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Violet 450)
Anticuerpos
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR