Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE (Phycoerythrin)/iFluor® 594)

Proveedor: AAT BIOQUEST
104411Y0 104411Y1 104411Y2
AATB104411Y0EA 319 EUR
AATB104411Y0 AATB104411Y1 AATB104411Y2
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE (Phycoerythrin)/iFluor® 594)
Anticuerpos
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: PE (Phycoerythrin)/iFluor® 594
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR