Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 450)

Proveedor: AAT BIOQUEST
10052040 10052041
AATB10052040EA 371 EUR
AATB10052040 AATB10052041
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 450)
Anticuerpos
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR