Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 514)
Proveedor: AAT BIOQUEST
10280060 10280061
AATB10280060EA 371 EUR
AATB10280060 AATB10280061
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 514)
Anticuerpos
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Precaución: For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 514
Epitope:
Host: Mouse
Isotype:
Reactivity: Human