Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 555)
Proveedor: AAT BIOQUEST
10052090 10052091
AATB10052090EA 371 EUR
AATB10052090 AATB10052091
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 555)
Anticuerpos
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Precaución: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 555
Epitope:
Host: Mouse
Isotype:
Reactivity: Human