Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Blue 570)

Proveedor: AAT BIOQUEST
116100T0 116100T1
AATB116100T0EA 371 EUR
AATB116100T0 AATB116100T1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Blue 570)
Anticuerpos
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR