Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Blue 570)

Proveedor: AAT BIOQUEST
102800T0 102800T1
AATB102800T0EA 371 EUR
AATB102800T0 AATB102800T1
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Blue 570)
Anticuerpos
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR