Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PE)

Proveedor: AAT Bioquest
101611L0 101611L1 101611L2
AATB101611L0EA 271 EUR
AATB101611L0 AATB101611L1 AATB101611L2
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PE)
Anticuerpos
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR