Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 647)

Proveedor: AAT BIOQUEST
100321Q0 100321Q1 100321Q2
AATB100321Q0EA 319 EUR
AATB100321Q0 AATB100321Q1 AATB100321Q2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 647)
Anticuerpos
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR