Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE/iFluor® 647)

Proveedor: AAT BIOQUEST
100811Q0 100811Q1 100811Q2
AATB100811Q0EA 319 EUR
AATB100811Q0 AATB100811Q1 AATB100811Q2
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE/iFluor® 647)
Anticuerpos
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PE/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR