Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 860)

Proveedor: AAT BIOQUEST
100410R0 100410R1
AATB100410R0EA 371 EUR
AATB100410R0 AATB100410R1
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 860)
Anticuerpos
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR