Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 810)

Proveedor: AAT BIOQUEST
101910O0 101910O1
AATB101910O0EA 371 EUR
AATB101910O0 AATB101910O1
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 810)
Anticuerpos
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 810
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno CD19
Nombre del antígeno CD19 antigen
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado iFluor® 810
Clon SJ25C1
Reactividad Human
Huésped Mouse
Isotipo
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Purification Affinity chromatography
Adsorción cruzada No
Temperatura de almacenamiento 2...8 °C with minimized light exposure. Do not freeze.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR