Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 820)
Proveedor: AAT BIOQUEST
100510P0 100510P1
AATB100510P0EA 371 EUR
AATB100510P0 AATB100510P1
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 820)
Anticuerpos
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Precaución: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 820
Epitope:
Host: Mouse
Isotype:
Reactivity: Human