Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (mFluor™ Green 620)

Proveedor: AAT BIOQUEST
100510U0 100510U1
AATB100510U0EA 371 EUR
AATB100510U0 AATB100510U1
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (mFluor™ Green 620)
Anticuerpos
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: mFluor™ Green 620
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR