Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ Violet 500)

Proveedor: AAT BIOQUEST
10041100 10041101
AATB10041100EA 371 EUR
AATB10041100 AATB10041101
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ Violet 500)
Anticuerpos
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Affinity purified
  • Easily reconstituted

Precaución: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno CD4
Environmentally Preferable
Nombre del antígeno CD4 antigen
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado mFluor™ Violet 500
Clon RPA-T4
Reactividad Human
Huésped Mouse
Isotipo IgG1
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Citometría de flujo Yes
Purification Affinity chromatography
Adsorción cruzada No
Temperatura de almacenamiento Freeze (<–15 °C); Minimise light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR