Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (Pacific Orange)

Proveedor: AAT BIOQUEST
100521K0 100521K1
AATB100521K0EA 371 EUR
AATB100521K0 AATB100521K1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (Pacific Orange)
Anticuerpos
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: Pacific Orange
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR