Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Proveedor: AAT BIOQUEST
102801U0 102801U1 102801U2
AATB102801U0EA 319 EUR
AATB102801U0 AATB102801U1 AATB102801U2
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Anticuerpos
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR