Bizflame™ Work Shirts
Proveedor: Portwest
FR69NARL FR69NARM FR69NARS FR69NARXL FR69NARXXL FR69NARXXXL
PRTWFR69NARLEA 58.1 EUR
PRTWFR69NARL PRTWFR69NARM PRTWFR69NARS PRTWFR69NARXL PRTWFR69NARXXL PRTWFR69NARXXXL
Bizflame™ Work Shirts
Ropa de trabajo
Offering a more formal look, this lightweight long sleeve shirt protects against heat, flame, anti-static, and light chemical hazards.
  • Protects against radiant and convective heat
  • Lightweight and comfortable
  • Chest pockets with stud closure
  • Concealed stud front for easy access
  • Stud adjustable cuffs for a secure fit
  • Shirt tail hem stays tucked in
  • Two pockets for secure storage
The concealed stud front fastening enhances the professional appearance while also offering maximum FR protection and all day comfort. Recommended for use in dry, indoor environments.
Certificaciones: EN ISO 11612 A1, B1, C2. EN 1149 -5. EN 13034 Type PB [6]. CE-CAT III.