Docosanoato de metilo, Supelco®
Proveedor: Merck
Sinónimos:
Methyl behenate, Docosanoic acid methyl ester, Behenic acid methyl ester
M-Clarity™ quality level = MQ100
11940-1G
11940-5G
11940-1GEA
69.5
EUR
11940-1G
11940-5G
Docosanoato de metilo, Supelco®
Docosanoato de metilo
Formula:
C₂₃H₄₆O₂ |
Número MDL:
MFCD00009347 Núm. CAS: 929-77-1 EINECS: 213-207-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...