Anti-CD11c Hamster (Armenian) Monoclonal Antibody [clone: N418] (PE)

Proveedor: STEMCELL Technologies
100-0445 100-0446
STMC100-0445EA 72 EUR
STMC100-0445 STMC100-0446
Anti-CD11c Hamster (Armenian) Monoclonal Antibody [clone: N418] (PE)
Anticuerpos
Hamster (Armenian) monoclonal IgG antibody against mouse CD11c.

  • Dendritic cells

The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor. Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis. In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780).

Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: PE
Epitope:
Host: Hamster (Armenian)
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR