Anti-CD11c Hamster Monoclonal Antibody [clone: N418] (Pacific Blue™)
Proveedor: STEMCELL Technologies
60002PB.1 60002PB
STMC60002PB.1EA 145 EUR
STMC60002PB.1 STMC60002PB
Anti-CD11c Hamster Monoclonal Antibody [clone: N418] (Pacific Blue™)
Anticuerpos
Hamster (Armenian) monoclonal IgG antibody against mouse CD11c, Pacific Blue™-conjugated.
  • Dendritic cells
The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor. Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cellMore Product Information