Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 430)
Proveedor: AAT BIOQUEST
50050 50051
AATB50050EA 153 EUR
AATB50050 AATB50051
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 430)
Anticuerpos
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
  • Affinity purified
  • Easily reconstituted
Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).
Precaución: For research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 430
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human