Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (XFD750)

Proveedor: AAT BIOQUEST
50182 50183
AATB50182EA 153 EUR
AATB50182 AATB50183
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (XFD750)
Anticuerpos
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Precaución: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: XFD750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR