Anti-E Cadherin Recombinant Recombinant Antibody [clone: DMC484]

Proveedor: ANTIBODIES.COM
A318706-100 A318706-500
ANTIA318706-100EA 640 EUR
ANTIA318706-100 ANTIA318706-500
Anti-E Cadherin Recombinant Recombinant Antibody [clone: DMC484]
Anticuerpos
Recombinant chimeric monoclonal [DMC484] antibody to E Cadherin for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: E Cadherin
Clonality: Recombinant
Clone: DMC484
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR