Anti-GFP Goat Polyclonal Antibody
Proveedor: ANTIBODIES.COM
A121560-1 A121560-400
ANTIA121560-1EA 635 EUR
ANTIA121560-1 ANTIA121560-400
Anti-GFP Goat Polyclonal Antibody
Anticuerpos
Goat polyclonal antibody to GFP for WB, IF, IHC-P and IHC-Fr with samples derived from GFP, YFP and CFP.
  • Validated applications: WB, IF, IHC-P, IHC to Fr
  • Recommended dilutions: WB: 1:500 to 1:5,000, IF: 1:50 to 1:1,000, IHC-P: 1:50 to 1:1,000, IHC to Fr: 1:50 to 1:1,000
Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: GFP, YFP, CFP