Anti-Calnexin Goat Polyclonal Antibody
Proveedor: ANTIBODIES.COM
A121570-1 A121570-400
ANTIA121570-1EA 635 EUR
ANTIA121570-1 ANTIA121570-400
Anti-Calnexin Goat Polyclonal Antibody
Anticuerpos
Goat polyclonal antibody to Calnexin for WB, IF, IHC-P and IHC-Fr with samples derived from human, rat, mouse, canine and monkey.
  • Validated applications: WB, IF, IHC-P, IHC to Fr
  • Recommended dilutions: WB: 1:500 to 1:5,000, IF: 1:50 to 1:500, IHC-P: 1:200 to 1:1,000, IHC to Fr: 1:200 to 1:1,000
Type: Primary
Antigen: Calnexin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Mouse, Dog, Monkey