Anti-ERp57 Goat Polyclonal Antibody

Proveedor: ANTIBODIES.COM
A121547-1 A121547-400
ANTIA121547-1EA 635 EUR
ANTIA121547-1 ANTIA121547-400
Anti-ERp57 Goat Polyclonal Antibody
Anticuerpos
Goat polyclonal antibody to ERp57 for WB, IF, IHC-P and IHC-Fr with samples derived from human, rat, mouse, canine and monkey.

  • Validated applications: WB, IF, IHC-P, IHC to Fr
  • Recommended dilutions: WB: 1:500 to 1:2000, IF: 1:50 to 1:500, IHC-P: 1:200 to 1:1000, IHC to Fr: 1:200 to 1:1000

Type: Primary
Antigen: ERp57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat;Mouse;Dog;Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR