Anti-mCherry Goat Polyclonal Antibody
Proveedor: ANTIBODIES.COM
A121571-15 A121571-600
ANTIA121571-15EA 635 EUR
ANTIA121571-15 ANTIA121571-600
Anti-mCherry Goat Polyclonal Antibody
Anticuerpos
Goat polyclonal antibody to mCherry for WB, IF, IHC-P, IHC-Fr and IEM with samples derived from mCherry, tdTomato and Red Fluorescent Protein (dsRed).
  • Validated applications: WB, IF, IHC-P, IHC to Fr, IEM
  • Recommended dilutions: WB: 1:500 to 1:5000, IF: 50 to 1:500, IHC-P: 1:50 to 1:500, IHC to Fr: 1:50 to 1:500
Type: Primary
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: mCherry;tdTomato;Red Fluorescent Protein (dsRed)