Anti-Collagen III Mouse Monoclonal Antibody [clone: ABT-CIII]

Proveedor: ANTIBODIES.COM
A95291-100 A95291-50
ANTIA95291-100EA 475 EUR
ANTIA95291-100 ANTIA95291-50
Anti-Collagen III Mouse Monoclonal Antibody [clone: ABT-CIII]
Anticuerpos
Mouse monoclonal (ABT-CIII) antibody to Collagen Type III for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Collagen III
Clonality: Monoclonal
Clone: ABT-CIII
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR