Anti-Coronin 1a Rabbit Polyclonal Antibody

Proveedor: ANTIBODIES.COM
A85431-100 A85431-50
ANTIA85431-100EA 520 EUR
ANTIA85431-100 ANTIA85431-50
Anti-Coronin 1a Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody to Coronin 1a for WB and ICC/IF with samples derived from Human, Bovine, Porcine, Rat and Mouse.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:5000, ICC/IF: 1:500 to 1:100

Type: Primary
Antigen: Coronin 1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Bovine;Porcine;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR