Anti-Pdi1p Mouse Monoclonal Antibody [clone: 38H8]

Proveedor: ANTIBODIES.COM
A85436-250 A85436-500
ANTIA85436-250EA 395 EUR
ANTIA85436-250 ANTIA85436-500
Anti-Pdi1p Mouse Monoclonal Antibody [clone: 38H8]
Anticuerpos
Mouse monoclonal (38H8) antibody to Pdi1p for WB, IF and IP with samples derived from Saccharomyces cerevisiae.

  • Validated applications: WB, IF, IP
  • Recommended dilutions: WB: 1:10000, IF: 1:500 to 1:100

Type: Primary
Antigen: Pdi1p
Clonality: Monoclonal
Clone: 38H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Saccharomyces cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR