Anti-NF-H Mouse Monoclonal Antibody [clone: AH1]

Proveedor: ANTIBODIES.COM
A85340-100 A85340-50
ANTIA85340-100EA 520 EUR
ANTIA85340-100 ANTIA85340-50
Anti-NF-H Mouse Monoclonal Antibody [clone: AH1]
Anticuerpos
Mouse monoclonal (AH1) antibody to NF-H for WB, ICC/IF, IHC-Fr and IHC-P with samples derived from Human, Rat, Mouse, Bovine, Porcine and Horse.

  • Validated applications: WB, ICC/IF, IHC to Fr, IHC to P
  • Recommended dilutions: WB: 1:10000, ICC/IF: 1:1000, IHC: 1:100

Type: Primary
Antigen: NF-H
Clonality: Monoclonal
Clone: AH1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse;Bovine;Porcine;Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR