Anti-NF-H Mouse Monoclonal Antibody [clone: 9B12]

Proveedor: ANTIBODIES.COM
A85341-100 A85341-50
ANTIA85341-100EA 520 EUR
ANTIA85341-100 ANTIA85341-50
Anti-NF-H Mouse Monoclonal Antibody [clone: 9B12]
Anticuerpos
Mouse monoclonal (9B12) antibody to NF-H for WB, ICC/IF, IHC-Fr and IHC-P with samples derived from Human, Rat, Mouse, Bovine and Chicken.

  • Validated applications: WB, ICC/IF, IHC to Fr, IHC to P
  • Recommended dilutions: WB: 1:10000, ICC/IF: 1:1000, IHC: 1:100

Type: Primary
Antigen: NF-H
Clonality: Monoclonal
Clone: 9B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Rat;Mouse;Bovine;Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR