CM-DiI, CytoTrace™
Proveedor: AAT BIOQUEST
22057 22058
AATB22057EA 202 EUR
AATB22057 AATB22058
CM-DiI, CytoTrace™
CM-DiI
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine iodide (CM-DiI) is a lipophilic carbocyanine dye that is commonly used for labeling cell membranes in various biological and neuroscientific research applications.
  • Stable for 12 months
Quality tested in-house
Precaución: For Research Use Only