Psoralen-A17-Biotin, ReadiLink™
Proveedor: AAT BIOQUEST
39052
39053
AATB39052EA
175
EUR
AATB39052
AATB39053
Psoralen-A17-Biotin, ReadiLink™
Psoralen-A17-Biotin
ReadiLink™ Psoralen-A17-Biotin is a photoactivatable reagent for biotinylating DNA or RNA.
- Stable for 24 months
Quality tested in-house
Precaución: For Research Use Only