Buccutite™ Streptavidin Antibody Conjugation Kit
Proveedor: AAT BIOQUEST
5511 5510 5509
AATB5511EA 673 EUR
AATB5511 AATB5510 AATB5509
Buccutite™ Streptavidin Antibody Conjugation Kit
Sistemas de enlace y etiquetado de proteínas
Buccutite™ Streptavidin antibody conjugation kits are optimised for labelling either 25 µg, 100 µg or 1 mg of protein.
  • The produced streptavidin-conjugated antibodies may be directly used in WB, ELISA, IHC without further purification
  • Can also be used to conjugate other proteins or peptides
  • It can be used to generate conjugates of different ratios of streptavidin/antibody
This streptavidin conjugation kit uses a simple and quick process for crosslinking streptavidin to an antibody.
Información de suministro: Shipped at RT.
Precaución: Research use only.