4-Amino-α,α,α-trifluoro-o-toluic acid
Proveedor: TCI
Sinónimos:
4-Amino-2-(trifluoromethyl)benzoic acid
A2889-25G
A2889-5G
TCIAA2889-25GEA
285
EUR
TCIAA2889-25G
TCIAA2889-5G
4-Amino-α,α,α-trifluoro-o-toluic acid
4-Amino-α,α,α-trifluoro-o-toluic acid
Formula:
C₈H₆F₃NO₂ MW: 205,14 g/mol |
Número MDL:
MFCD03407959 Núm. CAS: 393-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...