7-Bromo-1-heptene

Proveedor: TCI

B3255-1G B3255-5G
TCIAB3255-1GEA 30 EUR
TCIAB3255-1G TCIAB3255-5G
7-Bromo-1-heptene
7-Bromo-1-heptene
Formula: C₇H₁₃Br
Número MDL: MFCD01631137
Núm. CAS: 4117-09-3
UN: 1993

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR