1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected

Proveedor: TCI

Sinónimos: 1-(Tert-butoxycarbonyl)-5-(4,4,5,5-Tetramethyl-[1,3,2]Dioxaborolan-2-yl)indazole, 1-Boc-1H-indalzole-5-boronic acid pinacol ester, tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

B5992-1G B5992-200MG
TCIAB5992-1GEA 129 EUR
TCIAB5992-1G TCIAB5992-200MG
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
Formula: C₁₈H₂₅BN₂O₄
Número MDL: MFCD08437629
Núm. CAS: 864771-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR