N,N'-Bis(acryloyl)cystamine
Proveedor: TCI
Sinónimos:
N,N'-(Disulphanediylbis(ethane-2,1-diyl))diacrylamide
B6238-1G
B6238-5G
TCIAB6238-1GEA
46
EUR
TCIAB6238-1G
TCIAB6238-5G
N,N'-Bis(acryloyl)cystamine
N,N'-Bis(acryloyl)cystamine
Formula:
C₁₀H₁₆N₂O₂S₂ MW: 260,37 g/mol |
Número MDL:
MFCD00036225 Núm. CAS: 60984-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...