A-769662
Proveedor: TCI
Sinónimos:
4-Hydroxy-3-(2'-hydroxy-[1,1'-biphenyl]-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, A 769662
H1552-10MG
H1552-50MG
TCIAH1552-10MGEA
84
EUR
TCIAH1552-10MG
TCIAH1552-50MG
A-769662
A-769662
Formula:
C₂₀H₁₂N₂O₃S MW: 360,387 g/mol |
Número MDL:
MFCD11977269 Núm. CAS: 844499-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...