2-Iodo-4-methylphenol

Proveedor: TCI

I1132-1G I1132-5G
TCIAI1132-1GEA 39 EUR
TCIAI1132-1G TCIAI1132-5G
2-Iodo-4-methylphenol
2-Iodo-4-methylphenol
Formula: C₇H₇IO
MW: 234.03 g/mol
Número MDL: MFCD00189405
Núm. CAS: 16188-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR